Order Entry
Export
Orders LinkContactUsLinkComponent
Calmagite ≥98%

Calmagite ≥98%

Bottle
Catalog # APOSBUP18203-500MG
New Product
Supplier:   Apollo Scientific
Calmagite ≥98%
Catalog # APOSBUP18203-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  3147-14-6
Calmagite ≥98%

Bottle

CAS Number:  3147-14-6
Price
37,70 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

  • CAS Number:3147-14-6
  • Formula:C₁₇H₁₄N₂O₅S
  • Molecular Weight:358,37 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications