Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
cis-4-Hydroxy-L(-)-proline
BP0000733363
cis-4-Hydroxy-L-proline
Print
cis-4-Hydroxy-L-proline
New Product
Supplier:
Apollo Scientific
cis-4-Hydroxy-L-proline
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
cis-4-Hydroxy-L-proline
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP17950-200MG
Bottle
Log in
for availability
Log in
for availability
1 * 200 mg
1 * 200 mg - 42,00 €
42,00 €
-
+
1 * 200 mg
42,00 €
-
+
Specifications:
New Product
New Product
APOSBUP17950-500MG
Bottle
Log in
for availability
Log in
for availability
1 * 500 mg
1 * 500 mg - 71,00 €
71,00 €
-
+
1 * 500 mg
71,00 €
-
+
Specifications:
New Product
New Product
APOSBUP17950-1G
Bottle
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 102,00 €
102,00 €
-
+
1 * 1 g
102,00 €
-
+
Specifications:
New Product
New Product