Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Tributyl acetylcitrate
BP0000733292
Print
Acetyltributyl citrate
New Product
Supplier:
Apollo Scientific
Acetyltributyl citrate
Supplier:
Apollo Scientific
New Product
Certificates
Return to Top
Acetyltributyl citrate
Product Family Options
Jump to:
Product Family Options
About this item
2 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP17184-100MG
Bottle
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 316,00 €
316,00 €
-
+
1 * 100 mg
316,00 €
-
+
Specifications:
New Product
New Product
APOSBUP17184-200MG
Bottle
Log in
for availability
Log in
for availability
1 * 200 mg
1 * 200 mg - 469,00 €
469,00 €
-
+
1 * 200 mg
469,00 €
-
+
Specifications:
New Product
New Product