Order Entry
Export
Orders LinkContactUsLinkComponent
AC-PHE-OME ≥98%

AC-PHE-OME ≥98%

Bottle
Catalog # APOSBUP16675-100MG
New Product
Supplier:   Apollo Scientific
AC-PHE-OME ≥98%
Catalog # APOSBUP16675-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  3618-96-0
AC-PHE-OME ≥98%

Bottle

CAS Number:  3618-96-0
Price
39,10 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Methyl N-acetyl-L-phenylalaninate
  • CAS Number:3618-96-0
  • Formula:C₁₂H₁₅NO₃
  • Molecular Weight:221,25 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications