Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
4,7-Dihydroxycoumarin
BP0000733064
Print
4,7-DIHYDROXYCOUMARIN
New Product
Supplier:
Apollo Scientific
4,7-DIHYDROXYCOUMARIN
Supplier:
Apollo Scientific
New Product
Certificates
Return to Top
4,7-DIHYDROXYCOUMARIN
Product Family Options
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP14724-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 44,90 €
44,90 €
-
+
1 * 50 mg
44,90 €
-
+
Specifications:
New Product
New Product
APOSBUP14724-100MG
Bottle
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 68,10 €
68,10 €
-
+
1 * 100 mg
68,10 €
-
+
Specifications:
New Product
New Product
APOSBUP14724-500MG
Bottle
Log in
for availability
Log in
for availability
1 * 500 mg
1 * 500 mg - 164,00 €
164,00 €
-
+
1 * 500 mg
164,00 €
-
+
Specifications:
New Product
New Product