0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,10-Decanediol
BP0000730867
1,10-Decanediol
New Product
:
Apollo Scientific
1,10-Decanediol
:
Apollo Scientific
New Product
1,10-Decanediol
Product Family Options
Pack type
APOSOR93674-25G
Bottle
1 * 25 g
1 * 25 g - 21,80 €
21,80 €
-
+
1 * 25 g
21,80 €
-
+
New Product
New Product
APOSOR93674-100G
Bottle
1 * 100 g
1 * 100 g - 27,50 €
27,50 €
-
+
1 * 100 g
27,50 €
-
+
New Product
New Product
APOSOR93674-500G
Bottle
1 * 500 g
1 * 500 g - 81,10 €
81,10 €
-
+
1 * 500 g
81,10 €
-
+
New Product
New Product