Order Entry
Export
Orders LinkContactUsLinkComponent
2-Nitroethanol 98%

2-Nitroethanol 98%

Irritant
Supplier:   Apollo Scientific
2-Nitroethanol 98%
Supplier:  Apollo Scientific
Certificates
2-Nitroethanol 98%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:625-48-9
  • Formula:C₂H₅NO₃
  • MDL Number:MFCD00007405
  • Molecular Weight:91,065 g/mol
  • EINECS:210-895-1
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 98,40 €
    98,40 €
    1 * 25 g
       98,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 229,00 €
    229,00 €
    1 * 100 g
       229,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 130,00 €
    1 130,00 €
    1 * 500 g
       1 130,00 €

    Specifications:
    New Product
    New Product