4-Tritylphenol 97%
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
- CAS Number:978-86-9
- Formula:C₂₅H₂₀O
- MDL Number:MFCD00002364
- Molecular Weight:336,43 g/mol
- EINECS:213-557-1
5 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Number:978-86-9
- Formula:C₂₅H₂₀O
- MDL Number:MFCD00002364
- Molecular Weight:336,43 g/mol
- EINECS:213-557-1
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



