0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,8-Dibromodibenzofuran
BP0000729269
2,8-Dibromodibenzo[b,d]furan
2,8-Dibromodibenzo[b,d]furan
New Product
:
Apollo Scientific
2,8-Dibromodibenzo[b,d]furan
:
Apollo Scientific
New Product
Expand image
2,8-Dibromodibenzo[b,d]furan
Add to cart
Product Family Options
Pack type
APOSOR74699-250MG
Bottle
1 * 250 mg
1 * 250 mg - 21,80 €
21,80 €
-
+
1 * 250 mg
21,80 €
-
+
New Product
New Product
APOSOR74699-1G
Bottle
1 * 1 g
1 * 1 g - 24,60 €
24,60 €
-
+
1 * 1 g
24,60 €
-
+
New Product
New Product
APOSOR74699-5G
Bottle
1 * 5 g
1 * 5 g - 30,40 €
30,40 €
-
+
1 * 5 g
30,40 €
-
+
New Product
New Product
APOSOR74699-25G
Bottle
1 * 25 g
1 * 25 g - 161,00 €
161,00 €
-
+
1 * 25 g
161,00 €
-
+
New Product
New Product
APOSOR74699-100G
Bottle
1 * 100 g
1 * 100 g - 637,00 €
637,00 €
-
+
1 * 100 g
637,00 €
-
+
New Product
New Product
APOSOR74699-500G
Bottle
1 * 500 g
1 * 500 g - 1 770,00 €
1 770,00 €
-
+
1 * 500 g
1 770,00 €
-
+
New Product
New Product