0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
17-AAG
BP0000728799
Tanespimycin
Bottle
APOSBUP11464-25MG
New Product
:
Apollo Scientific
Tanespimycin
APOSBUP11464-25MG
:
Apollo Scientific
New Product
:
Tanespimycin
Bottle
:
125,00 €
1 * 25 mg
-
+
Pack type:
Bottle