TES-1025 ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/life science applications
Synonyms: TES-1025
- CAS Number:1883602-21-8
- Formula:C₁₈H₁₃N₃O₃S₂
- Molecular Weight:383,44 g/mol
3 Options Available Below
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: TES-1025
- CAS Number:1883602-21-8
- Formula:C₁₈H₁₃N₃O₃S₂
- Molecular Weight:383,44 g/mol



