Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
BAY 41-2272
BP0000728653
BAY 41-2272
Print
BAY 41-2272
New Product
Supplier:
Apollo Scientific
BAY 41-2272
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
BAY 41-2272
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP09895-25MG
Bottle
Log in
for availability
Log in
for availability
1 * 25 mg
1 * 25 mg - 220,00 €
220,00 €
-
+
1 * 25 mg
220,00 €
-
+
Specifications:
New Product
New Product
APOSBUP09895-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 349,00 €
349,00 €
-
+
1 * 50 mg
349,00 €
-
+
Specifications:
New Product
New Product
APOSBUP09895-100MG
Bottle
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 503,00 €
503,00 €
-
+
1 * 100 mg
503,00 €
-
+
Specifications:
New Product
New Product