Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
WP-1130
BP0000728391
Print
Degrasyn
New Product
Supplier:
Apollo Scientific
Degrasyn
Supplier:
Apollo Scientific
New Product
Certificates
Return to Top
Degrasyn
Product Family Options
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP06025-25MG
Bottle
Log in
for availability
Log in
for availability
1 * 25 mg
1 * 25 mg - 267,00 €
267,00 €
-
+
1 * 25 mg
267,00 €
-
+
Specifications:
New Product
New Product
APOSBUP06025-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 479,00 €
479,00 €
-
+
1 * 50 mg
479,00 €
-
+
Specifications:
New Product
New Product
APOSBUP06025-100MG
Bottle
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 711,00 €
711,00 €
-
+
1 * 100 mg
711,00 €
-
+
Specifications:
New Product
New Product