Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
N,N'-(9-((4-(Dimethylamino)phenyl)amino)acridine-3,6-diyl)bis(3-(pyrrolidin-1-yl)propanamide)
BP0000728216
Braco-19
Print
Braco-19
New Product
Supplier:
Apollo Scientific
Braco-19
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
Braco-19
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Availability
Price
APOSBUP03340-25MG
Log in
for availability
Log in
for availability
1 * 25 mg
1 * 25 mg - 526,00 €
526,00 €
-
+
1 * 25 mg
526,00 €
-
+
Specifications:
New Product
New Product
APOSBUP03340-50MG
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 837,00 €
837,00 €
-
+
1 * 50 mg
837,00 €
-
+
Specifications:
New Product
New Product
APOSBUP03340-100MG
Log in
for availability
Log in
for availability
1 * 100 mg
1 * 100 mg - 1 200,00 €
1 200,00 €
-
+
1 * 100 mg
1 200,00 €
-
+
Specifications:
New Product
New Product