Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Benzoic acid
BP0000728012
Benzoic acid
Print
Benzoic acid
Catalog # APOSBIA141014-1KG
New Product
Supplier:
Apollo Scientific
Benzoic acid
Catalog # APOSBIA141014-1KG
Supplier:
Apollo Scientific
New Product
CAS Number:
Expand image
Return to Top
Benzoic acid
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
181,00 €
Log in
for availability
Unit of Measure
1 * 1 kg
-
+
Add to cart
Return Policy
Product Details & Documents
Specifications
Specifications
UN No.:
2811
Cat. No.:
1517006.
Description:
Phenelzine Sulfate
CAS No.:
156-51-4
Specifications