Order Entry
Export
Orders LinkContactUsLinkComponent
3-Hydroxyazetidine 98%

3-Hydroxyazetidine 98%

Irritant
Supplier:   Apollo Scientific
3-Hydroxyazetidine 98%
Supplier:  Apollo Scientific
Certificates
3-Hydroxyazetidine 98%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  3-Azetidinol, Azetidin-3-ol
  • CAS Number:45347-82-8
  • Formula:C₃H₇NO
  • MDL Number:MFCD03695446
  • Molecular Weight:73,09377 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 110,00 €
    110,00 €
    1 * 1 g
       110,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 490,00 €
    490,00 €
    1 * 5 g
       490,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 783,00 €
    783,00 €
    1 * 10 g
       783,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 570,00 €
    1 570,00 €
    1 * 25 g
       1 570,00 €