Order Entry
Export
Orders LinkContactUsLinkComponent
 

Perylene 98%

Irritant
Supplier:   Apollo Scientific
Perylene 98%
Supplier:  Apollo Scientific
Certificates

5 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:198-55-0
  • Formula:C₂₀H₁₂
  • MDL Number:MFCD00004142
  • Molecular Weight:252,31 g/mol
  • EINECS:205-900-9
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 69,50 €
    69,50 €
    1 * 25 g
       69,50 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 199,00 €
    199,00 €
    1 * 100 g
       199,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 974,00 €
    974,00 €
    1 * 500 g
       974,00 €

    Specifications:
    New Product
    New Product