0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
DL-Malic acid
BP0000726352
Malic acid
New Product
:
Apollo Scientific
Malic acid
:
Apollo Scientific
New Product
Malic acid
Product Family Options
Pack type
APOSOR1013709-100G
Bottle
1 * 100 g
1 * 100 g - 21,80 €
21,80 €
-
+
1 * 100 g
21,80 €
-
+
New Product
New Product
APOSOR1013709-500G
Bottle
1 * 500 g
1 * 500 g - 57,90 €
57,90 €
-
+
1 * 500 g
57,90 €
-
+
New Product
New Product
APOSOR10137092.5KG
Bottle
1 * 2,5 kg
1 * 2,5 kg - 261,00 €
261,00 €
-
+
1 * 2,5 kg
261,00 €
-
+
New Product
New Product
APOSOR1013709-25KG
Bottle
1 * 25 kg
1 * 25 kg - 2 540,00 €
2 540,00 €
-
+
1 * 25 kg
2 540,00 €
-
+
New Product
New Product