Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl Gallate 98%

Ethyl Gallate 98%

Supplier:   Apollo Scientific
Ethyl Gallate 98%
Supplier:  Apollo Scientific
Certificates
Ethyl Gallate 98%

6 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate
  • CAS Number:831-61-8
  • Formula:C₉H₁₀O₅
  • MDL Number:MFCD00016430
  • Molecular Weight:198,17 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 24,60 €
    24,60 €
    1 * 25 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 66,60 €
    66,60 €
    1 * 100 g
       66,60 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 270,00 €
    270,00 €
    1 * 500 g
       270,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 953,00 €
    953,00 €
    1 * 2,5 kg
       953,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 3 720,00 €
    3 720,00 €
    1 * 10 kg
       3 720,00 €

    Specifications:
    New Product
    New Product