Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl Blue

Methyl Blue

Bottle
Catalog # APOSBUP18835-500MG
New Product
Supplier:   Apollo Scientific
Methyl Blue
Catalog # APOSBUP18835-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  28983-56-4
Methyl Blue

Bottle

CAS Number:  28983-56-4
Price
37,70 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Aniline blue sodium salt, [[4-[bis[4-[(sulphophenyl)amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]amino]-benzenesulphonic acid sodium salt (1:2), Poirriers blue, Cotton blue, Water blue, C.I. 42780, Aniline blue, water soluble, Disodium [[4-[bis[4-[(sulphonatophenyl)amino]phenyl]methylene]cyclohexa-2,5-dien-1-ylidene]amino]benzenesulphonate, Acid Blue 93
  • CAS Number:28983-56-4
  • Formula:C₃₇H₂₇N₃Na₂O₉S₃
  • Molecular Weight:799,8 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications