Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Miconazole
BP0000725427
Miconazole
Print
Miconazole
Bottle
Catalog # APOSBUP18727-500MG
New Product
Supplier:
Apollo Scientific
Miconazole
Catalog # APOSBUP18727-500MG
Supplier:
Apollo Scientific
New Product
CAS Number:
Expand image
Return to Top
Miconazole
Bottle
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
50,70 €
Log in
for availability
Unit of Measure
1 * 500 mg
-
+
Add to cart
Return Policy
Product Details & Documents
Specifications
Specifications
Pack type:
Bottle
Specifications