0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Chloramphenicol
BP0000725412
Chloramphenicol
New Product
:
Apollo Scientific
Chloramphenicol
:
Apollo Scientific
New Product
Chloramphenicol
Product Family Options
Pack type
APOSBUP18539-500MG
Bottle
1 * 500 mg
1 * 500 mg - 52,10 €
52,10 €
-
+
1 * 500 mg
52,10 €
-
+
New Product
New Product
APOSBUP18539-1G
Bottle
1 * 1 g
1 * 1 g - 62,30 €
62,30 €
-
+
1 * 1 g
62,30 €
-
+
New Product
New Product
APOSBUP18539-5G
Bottle
1 * 5 g
1 * 5 g - 82,50 €
82,50 €
-
+
1 * 5 g
82,50 €
-
+
New Product
New Product
APOSBUP18539-10G
Bottle
1 * 10 g
1 * 10 g - 119,00 €
119,00 €
-
+
1 * 10 g
119,00 €
-
+
New Product
New Product