Order Entry
Export
Orders LinkContactUsLinkComponent
3-Methylcatechol ≥90%

3-Methylcatechol ≥90%

New Product
Supplier:   Apollo Scientific
3-Methylcatechol ≥90%
Supplier:  Apollo Scientific
New Product
Certificates
3-Methylcatechol ≥90%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  3-Methylcatechol, 3-Methylbenzene-1,2-diol
  • CAS Number:488-17-5
  • Formula:C₇H₈O₂
  • Molecular Weight:124,14 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 40,60 €
    40,60 €
    1 * 500 mg
       40,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 55,00 €
    55,00 €
    1 * 1 g
       55,00 €

    Specifications:
    New Product
    New Product