0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,7-Diaminoheptane
BP0000725355
1,7-Diaminoheptane
1,7-Diaminoheptane
Bottle
APOSBUP17879-200MG
New Product
:
Apollo Scientific
1,7-Diaminoheptane
APOSBUP17879-200MG
:
Apollo Scientific
New Product
:
Expand image
1,7-Diaminoheptane
Bottle
Add to cart
:
63,70 €
Unit of Measure
1 * 200 mg
-
+
Add to cart
Pack type:
Bottle