Order Entry
Export
Orders LinkContactUsLinkComponent
4-POBN ≥98%

4-POBN ≥98%

New Product
Supplier:   Apollo Scientific
4-POBN ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  4-Aminobenzoic hydrazide, 4-Aminobenzene-1-carbohydrazide, 4-Aminobenzoic acid hydrazide, 4-Aminobenzohydrazide
  • CAS Number:5351-17-7
  • Formula:C₇H₉N₃O
  • Molecular Weight:151,17 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 43,50 €
    43,50 €
    1 * 200 mg
       43,50 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 68,10 €
    68,10 €
    1 * 500 mg
       68,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 98,40 €
    98,40 €
    1 * 1 g
       98,40 €

    Specifications:
    New Product
    New Product