Order Entry
Export
Orders LinkContactUsLinkComponent
Cetyldimethylethylammonium Bromide ≥98%

Cetyldimethylethylammonium Bromide ≥98%

Bottle
Catalog # APOSBUP17178-5G
New Product
Supplier:   Apollo Scientific
Cetyldimethylethylammonium Bromide ≥98%
Catalog # APOSBUP17178-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  124-03-8
Cetyldimethylethylammonium Bromide ≥98%

Bottle

CAS Number:  124-03-8
Price
4 120,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Ethylhexadecyldimethylammonium Bromide, Hexadecylethyldimethylammonium bromide, Ethyl(1-hexadecyl)dimethylammonium bromide, Ethylhexadecyldimethylammonium bromide, Dimethylethylhexadecylammonium bromide
  • CAS Number:124-03-8
  • Formula:C₂₀H₄₄BrN
  • Molecular Weight:378,47 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications