Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
TW-37
BP0000724336
Print
TW-37
New Product
Supplier:
Apollo Scientific
TW-37
Supplier:
Apollo Scientific
New Product
Certificates
Return to Top
TW-37
Product Family Options
Jump to:
Product Family Options
About this item
2 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP05404-25MG
Bottle
Log in
for availability
Log in
for availability
1 * 25 mg
1 * 25 mg - 287,00 €
287,00 €
-
+
1 * 25 mg
287,00 €
-
+
Specifications:
New Product
New Product
APOSBUP05404-50MG
Bottle
Log in
for availability
Log in
for availability
1 * 50 mg
1 * 50 mg - 474,00 €
474,00 €
-
+
1 * 50 mg
474,00 €
-
+
Specifications:
New Product
New Product