Arsenazo III indicator
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
Synonyms: 2,7-Bis(2-arsonophenylazo)chromotropic acid, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid, Arsenazo Ⅲ
- CAS Number:1668-00-4
- Formula:C₂₂H₁₈As₂N₄O₁₄S₂
- MDL Number:MFCD00036695
- Molecular Weight:776,374 g/mol
5 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: 2,7-Bis(2-arsonophenylazo)chromotropic acid, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid, Arsenazo Ⅲ
- CAS Number:1668-00-4
- Formula:C₂₂H₁₈As₂N₄O₁₄S₂
- MDL Number:MFCD00036695
- Molecular Weight:776,374 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



