1,9-Dichlorononane 98%
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
- CAS Number:821-99-8
- Formula:C₉H₁₈Cl₂
- MDL Number:MFCD00001024
- Molecular Weight:197,15 g/mol
5 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Number:821-99-8
- Formula:C₉H₁₈Cl₂
- MDL Number:MFCD00001024
- Molecular Weight:197,15 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



