Order Entry
Export
Orders LinkContactUsLinkComponent
Benzophenoneoxime 98%

Benzophenoneoxime 98%

New Product
Supplier:   Apollo Scientific
Benzophenoneoxime 98%
Supplier:  Apollo Scientific
New Product
Certificates
Benzophenoneoxime 98%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:574-66-3
  • Formula:C₁₃H₁₁NO
  • MDL Number:MFCD00051461
  • Molecular Weight:197,23 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 36,20 €
    36,20 €
    1 * 25 g
       36,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 129,00 €
    129,00 €
    1 * 100 g
       129,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 686,00 €
    686,00 €
    1 * 500 g
       686,00 €

    Specifications:
    New Product
    New Product