Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dimethoxybenzoic acid 98%

2,4-Dimethoxybenzoic acid 98%

Supplier:   Apollo Scientific
2,4-Dimethoxybenzoic acid 98%
Supplier:  Apollo Scientific
Certificates
2,4-Dimethoxybenzoic acid 98%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:91-52-1
  • Formula:C₉H₁₀O₄
  • MDL Number:MFCD00002434
  • Molecular Weight:182,1733 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 24,60 €
    24,60 €
    1 * 25 g
       24,60 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 72,40 €
    72,40 €
    1 * 100 g
       72,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 355,00 €
    355,00 €
    1 * 500 g
       355,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 760,00 €
    1 760,00 €
    1 * 2,5 kg
       1 760,00 €

    Specifications:
    New Product
    New Product