Lanosterol 60%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
Synonyms: (3S,5R,10S,13R,14R,17R)-4,4,10,13,14-Pentamethyl-17-((R)-6-methylhept-5-en-2-yl)-2,3,4,5,6,7,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol, Cryptosterol
- CAS Number:79-63-0
- Formula:C₃₀H₅₀O
- Molecular Weight:426,72 g/mol
5 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: (3S,5R,10S,13R,14R,17R)-4,4,10,13,14-Pentamethyl-17-((R)-6-methylhept-5-en-2-yl)-2,3,4,5,6,7,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol, Cryptosterol
- CAS Number:79-63-0
- Formula:C₃₀H₅₀O
- Molecular Weight:426,72 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



