Calmagite indicator
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
- CAS Number:3147-14-6
- Formula:C₁₇H₁₄N₂O₅S
- Molecular Weight:358,37 g/mol
5 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Number:3147-14-6
- Formula:C₁₇H₁₄N₂O₅S
- Molecular Weight:358,37 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



