Order Entry
Export
Orders LinkContactUsLinkComponent
 

Nonadecan-10-o 98%

New Product
Supplier:   Apollo Scientific
Nonadecan-10-o 98%
Supplier:  Apollo Scientific
New Product
Certificates
Nonadecan-10-o 98%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Nonadecan-10-ol, 10-Nonadecyl alcohol, 10-Hydroxy nonadecane
  • CAS Number:16840-84-9
  • Formula:C₁₉H₄₀O
  • MDL Number:MFCD00968670
  • Molecular Weight:284,52 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 26,10 €
    26,10 €
    1 * 100 mg
       26,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 56,50 €
    56,50 €
    1 * 250 mg
       56,50 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 141,00 €
    141,00 €
    1 * 1 g
       141,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 491,00 €
    491,00 €
    1 * 5 g
       491,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 2 650,00 €
    2 650,00 €
    1 * 25 g
       2 650,00 €

    Specifications:
    New Product
    New Product