Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead (II) Bromide 99,9%

Irritant
Environment
Health
Supplier:   Apollo Scientific
Lead (II) Bromide 99,9%
Supplier:  Apollo Scientific
Certificates
Lead (II) Bromide 99,9%

2 Options Available Below

Product Details & Documents

Usage: Inorganic salt or catalyst.

Synonyms:  Lead dibromide, Lead(II) bromide
  • CAS Number:10031-22-8
  • Formula:Pb·2Br
  • MDL Number:MFCD00011156
  • Molecular Weight:367,008 g/mol
  • EINECS:233-084-4
  • UN:2291
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 138,00 €
    138,00 €
    1 * 100 g
       138,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 319,00 €
    319,00 €
    1 * 500 g
       319,00 €