Orders LinkContactUsLinkComponent
Solvent Blue 38
New Product
Solvent Blue 38
New Product
Solvent Blue 38
Usage: Inorganic salt or catalyst.
 :  Direct Blue 86, C.I. 74180, Luxol® Fast Blue MBSN
  •  :1328-51-4
  •  :C₃₂H₁₂CuN₈Na₂O₆S₂
  •  :778,14 g/mol
Product Family Options
  • Pack type
  • Bottle
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €
    New Product
    New Product
  • Bottle
    1 * 5 g
    1 * 5 g - 23,20 €
    23,20 €
    1 * 5 g
       23,20 €
    New Product
    New Product
  • Bottle
    1 * 25 g
    1 * 25 g - 49,20 €
    49,20 €
    1 * 25 g
       49,20 €
    New Product
    New Product
  • Bottle
    1 * 100 g
    1 * 100 g - 109,00 €
    109,00 €
    1 * 100 g
       109,00 €
    New Product
    New Product
  • Bottle
    1 * 500 g
    1 * 500 g - 300,00 €
    300,00 €
    1 * 500 g
       300,00 €
    New Product
    New Product
  • Bottle
    1 * 2,5 kg
    1 * 2,5 kg - 1 380,00 €
    1 380,00 €
    1 * 2,5 kg
       1 380,00 €