Order Entry
Export
Orders LinkContactUsLinkComponent
 

α-Lipoic acid ≥98%

Bottle
Catalog # APOSBUP19692-1G
New Product
Supplier:   Apollo Scientific
α-Lipoic acid ≥98%
Catalog # APOSBUP19692-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1077-28-7
α-Lipoic acid ≥98%

Bottle

CAS Number:  1077-28-7
Price
37,70 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
  • CAS Number:1077-28-7
  • Formula:C₈H₁₄O₂S₂
  • Molecular Weight:206,33 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications