Order Entry
Export
Orders LinkContactUsLinkComponent
TMB ≥98%

TMB ≥98%

Bottle
Catalog # APOSBUP19250-1ML
New Product
Supplier:   Apollo Scientific
TMB ≥98%
Catalog # APOSBUP19250-1ML
Supplier:  Apollo Scientific
New Product
CAS Number:  54827-17-7
TMB ≥98%

Bottle

CAS Number:  54827-17-7
Price
39,10 €
Log in for availability
1 * 1 mL
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • Molecular Weight:240,34 g/mol
Specifications

Specifications

  • Pack type:
    Bottle
  • Cat. No.:
    APOSBUP19250-1ML

Specifications