Order Entry
Export
Orders LinkContactUsLinkComponent
Mandelic acid ≥98%

Mandelic acid ≥98%

Bottle
Catalog # APOSBUP19066-1G
New Product
Supplier:   Apollo Scientific
Mandelic acid ≥98%
Catalog # APOSBUP19066-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  90-64-2
Mandelic acid ≥98%

Bottle

CAS Number:  90-64-2
Price
39,10 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
  • CAS Number:90-64-2
  • Formula:C₈H₈O₃
  • Molecular Weight:152,15 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications