Order Entry
Export
Orders LinkContactUsLinkComponent
L-Cystine ≥98%

L-Cystine ≥98%

New Product
Supplier:   Apollo Scientific
L-Cystine ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
L-Cystine ≥98%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
  • CAS Number:56-89-3
  • Formula:C₆H₁₂N₂O₄S₂
  • Molecular Weight:240,3 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 52,10 €
    52,10 €
    1 * 500 mg
       52,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 62,30 €
    62,30 €
    1 * 1 g
       62,30 €

    Specifications:
    New Product
    New Product