Order Entry
Export
Orders LinkContactUsLinkComponent
D-Alanine ≥98%

D-Alanine ≥98%

Bottle
Catalog # APOSBUP18199-500MG
New Product
Supplier:   Apollo Scientific
D-Alanine ≥98%
Catalog # APOSBUP18199-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  338-69-2
D-Alanine ≥98%

Bottle

CAS Number:  338-69-2
Price
52,10 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
  • CAS Number:338-69-2
  • Formula:C₃H₇NO₂
  • Molecular Weight:89,09 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications