Order Entry
Export
Orders LinkContactUsLinkComponent
Nicotinamide ≥98%

Nicotinamide ≥98%

Bottle
Catalog # APOSBUP16764-10G
New Product
Supplier:   Apollo Scientific
Nicotinamide ≥98%
Catalog # APOSBUP16764-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  98-92-0
Nicotinamide ≥98%

Bottle

CAS Number:  98-92-0
Price
280,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
  • CAS Number:98-92-0
  • Formula:C₆H₆N₂O
  • Molecular Weight:122,12 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications