Order Entry
Export
Orders LinkContactUsLinkComponent
 

PD98059 ≥98%

New Product
Supplier:   Apollo Scientific
PD98059 ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one
  • CAS Number:167869-21-8
  • Formula:C₁₆H₁₃NO₃
  • Molecular Weight:267,28 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 138,00 €
    138,00 €
    1 * 50 mg
       138,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 239,00 €
    239,00 €
    1 * 100 mg
       239,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 355,00 €
    355,00 €
    1 * 200 mg
       355,00 €

    Specifications:
    New Product
    New Product