Order Entry
Export
Orders LinkContactUsLinkComponent
AP 18 ≥90%

AP 18 ≥90%

New Product
Supplier:   Apollo Scientific
AP 18 ≥90%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  4-(4-Chlorophenyl)-3-methylbut-3-en-2-oxime.
  • CAS Number:55224-94-7
  • Formula:C₁₁H₁₂ClNO
  • Molecular Weight:209,67 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 183,00 €
    183,00 €
    1 * 25 mg
       183,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 288,00 €
    288,00 €
    1 * 50 mg
       288,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 452,00 €
    452,00 €
    1 * 100 mg
       452,00 €

    Specifications:
    New Product
    New Product