2-Nitroethanol 98%
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent.
- CAS Number:625-48-9
- Formula:C₂H₅NO₃
- MDL Number:MFCD00007405
- Molecular Weight:91,066 g/mol
- EINECS:210-895-1
5 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent.
- CAS Number:625-48-9
- Formula:C₂H₅NO₃
- MDL Number:MFCD00007405
- Molecular Weight:91,066 g/mol
- EINECS:210-895-1
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



