Order Entry
Export
Orders LinkContactUsLinkComponent
 

beta-Citronellol 95%

Irritant
Supplier:   Apollo Scientific
beta-Citronellol 95%
Supplier:  Apollo Scientific
Certificates
beta-Citronellol 95%

3 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol
  • CAS Number:106-22-9
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00002935
  • Molecular Weight:156,2652 g/mol
  • EINECS:203-375-0
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 50,70 €
    50,70 €
    1 * 100 g
       50,70 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 81,10 €
    81,10 €
    1 * 500 g
       81,10 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 158,00 €
    158,00 €
    1 * 1 kg
       158,00 €

    Specifications:
    New Product
    New Product