Order Entry
Export
Orders LinkContactUsLinkComponent
3'-Acetylacetophenone 98%

3'-Acetylacetophenone 98%

Supplier:   Apollo Scientific
3'-Acetylacetophenone 98%
Supplier:  Apollo Scientific
Certificates
3'-Acetylacetophenone 98%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Benzene-1,3-bis(acetyl)
  • CAS Number:6781-42-6
  • Formula:C₁₀H₁₀O₂
  • MDL Number:MFCD00008740
  • Molecular Weight:162,18519 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 29,00 €
    29,00 €
    1 * 5 g
       29,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 91,20 €
    91,20 €
    1 * 25 g
       91,20 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 359,00 €
    359,00 €
    1 * 100 g
       359,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 770,00 €
    1 770,00 €
    1 * 500 g
       1 770,00 €

    Specifications:
    New Product
    New Product