Order Entry
Export
Orders LinkContactUsLinkComponent
LM22B-10 ≥98%

LM22B-10 ≥98%

New Product
Supplier:   Apollo Scientific
LM22B-10 ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  LM22B-10
  • CAS Number:342777-54-2
  • Formula:C₂₇H₃₃ClN₂O₄
  • Molecular Weight:485,01 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 378,00 €
    378,00 €
    1 * 25 mg
       378,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 597,00 €
    597,00 €
    1 * 50 mg
       597,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 948,00 €
    948,00 €
    1 * 100 mg
       948,00 €

    Specifications:
    New Product
    New Product