Antitubercular agent-41 ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/ life science applications.
Synonyms: [4-(4-fluorophenyl)-1-piperazinyl](6-methoxyfuro[2,3-b]quinolin-2-yl)-methanone
- CAS Number:901032-23-3
- Formula:C₂₃H₂₀FN₃O₃
- Molecular Weight:405,42 g/mol
4 Options Available Below
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: [4-(4-fluorophenyl)-1-piperazinyl](6-methoxyfuro[2,3-b]quinolin-2-yl)-methanone
- CAS Number:901032-23-3
- Formula:C₂₃H₂₀FN₃O₃
- Molecular Weight:405,42 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



