Order Entry
Export
Orders LinkContactUsLinkComponent
 

Xanthorrhizol ≥98%

New Product
Supplier:   Apollo Scientific
Xanthorrhizol ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Xanthorrhizol ≥98%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

  • CAS Number:30199-26-9
  • Formula:C₁₅H₂₂O
  • Molecular Weight:218,33 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 890,00 €
    1 890,00 €
    1 * 25 mg
       1 890,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 2 530,00 €
    2 530,00 €
    1 * 50 mg
       2 530,00 €

    Specifications:
    New Product
    New Product